missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17661-1-AP-150UL
This item is not returnable.
View return policy
Description
MFAP4 is a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region.Specifications
| MFAP4 | |
| Polyclonal | |
| Unconjugated | |
| MFAP4 | |
| MFAP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 287382, 4239, 76293 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55083, Q9D1H9 | |
| Mfap4 | |
| MFAP4 Fusion Protein Ag11920 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction