missing translation for 'onlineSavingsMsg'
Learn More

MFAP5, Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse Polyclonal Antibody

Brand:  Abnova H00008076-B01.50uL

Product Code. 16158185

  • € 349.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MSLLGPKVLLFLAAFIITSDWIPLGVNSQRGDDVTQATPETFTEDPNLVNDPATDETVLAVLADIAPSTDDLASLSEKNTTAECWDEKFTCTRLYSVHRPVKQCIHQLCFTSLRRMYIVNKEICSRLVCKEHEAMKDELCRQMAGLPPRRLRRSNYFRLPPCENVDLQRPNGL
Specifications

Specifications

MFAP5
Polyclonal
No additive
NM_003480.2
MFAP5
MFAP5 (NP_003471.1, 1 a.a. ∼ 173 a.a) full-length human protein.
RUO
8076
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
microfibrillar associated protein 5
MAGP2/MP25
Mouse
50 μL
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.