missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFF Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18439DL594
This item is not returnable.
View return policy
Description
MFF Polyclonal antibody specifically detects MFF in Human samples. It is validated for Western Blot
Specifications
| MFF | |
| Polyclonal | |
| Western Blot | |
| mitochondrial fission factor | |
| Recognizes region between residues Between Between 100 and 150.. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Endocrinology | |
| 56947 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction