missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFG-E8 Antibody (SPM291), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11482PE
This item is not returnable.
View return policy
Description
MFG-E8 Monoclonal antibody specifically detects MFG-E8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MFG-E8 | |
| Monoclonal | |
| PE | |
| BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, medin, MFG1, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
| Human milk fat globule membrane preparation (Uniprot: Q08431) | |
| 0.1 mL | |
| Cancer | |
| 4240 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SPM291 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction