Learn More
MFN1 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802196S
Encompass_Preferred
 View more versions of this product
Product Code. 17570723
  • € 242.00 / 30µL
Description
MFN1 Monoclonal antibody specifically detects MFN1 in Human samples. It is validated for Western Blot
Specifications
Specifications
MFN1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01730
Mouse
Affinity chromatography
RUO
920
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2E4
Unconjugated
CD4
2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1
Full length human recombinant protein of human CD4 produced in HEK293T cell.
30 μL