missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MFN2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA572811

Product Code. 15981465

  • € 528.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MFN2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MFN2
Polyclonal
Unconjugated
MFN2
CMT2A; CMT2A2; CPRP 1; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; hypertension-related protein 1; Kiaa0214; LOW QUALITY PROTEIN: mitofusin-2; Marf; MFN 2; MFN2; mg:cb01g09; Mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; mitofusin 2; Mitofusin2; mitofusin-2; si:dkeyp-104h9.2; Transmembrane GTPase MFN2; wu:fb79a11
Rabbit
Antigen affinity chromatography
RUO
170731, 64476, 9927
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95140, Q80U63, Q8R500
MFN2
MFN2 antibody for a 17 amino acid peptide near the center of human MFN2. The immunogen is within amino acids 250 - 300 of MFN2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.