missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFRP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578630
This item is not returnable.
View return policy
Description
Positive Control: HeLa Predicted Reactivity: Bovine (81%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MFRP is a member of the frizzled-related proteins. It may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen.
Specifications
| MFRP | |
| Polyclonal | |
| Unconjugated | |
| MFRP | |
| C1QTNF5; DKFZp586B0621; FLJ30570; MCOP5; Membrane frizzled-related protein; membrane-type frizzled-related protein; MFRP; MGC32938; NNO2; rd6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83552 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.07 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9BY79 | |
| MFRP | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human MFRP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction