missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00
Specifications
| Antigen | MFSD1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MFSD1 Polyclonal specifically detects MFSD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MFSD1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ14153, major facilitator superfamily domain containing 1, major facilitator superfamily domain-containing protein 1, SMAP4, SMAP-4, Smooth muscle cell-associated protein 4, UG0581B09 | |
| MFSD1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 64747 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DQRAERILHKEQGKTGEVIKLTDVKDFS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title