missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFSD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569623
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
MFSD1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
Specifications
| MFSD1 | |
| Polyclonal | |
| Unconjugated | |
| MFSD1 | |
| 1200003O06Rik; hypothetical protein LOC641486; major facilitator superfamily domain containing 1; major facilitator superfamily domain-containing protein 1; Major facilitator superfamily domain-containing protein 1-like protein; Mfsd1; mfsd1a; RGD1305751; SMAP4; SMAP-4; Smooth muscle cell-associated protein 4; UG0581B09; zgc:123187 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64747 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H3U5 | |
| MFSD1 | |
| synthetic peptide directed towards the middle region of human MFSD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction