Learn More
MFSD2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30555-1-AP-20UL
 View more versions of this product
Product Code. 18025515
  • € 145.00 / 20µL
Description
The MFSD2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MFSD2. This antibody recognizes human, mouse, rat antigen. The MFSD2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
MFSD2
400 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
FLJ14490, FLJ35904, FP1147, HMFN0656, MFSD2, MFSD2A, PP10484, PP9177, RP3 342P20.1, UNQ300/PRO341
20 μL
84879
Store at -20°C. Stable for one year after shipment.