missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD2B Polyclonal Antibody

MFSD2B Polyclonal Antibody
Brand: Invitrogen PA521050
This item is not returnable.
View return policy
Description
MFSD2B Polyclonal Antibody for Western Blot
Specifications
| MFSD2B | |
| Polyclonal | |
| Unconjugated | |
| MFSD2B | |
| hMfsd2b; major facilitator superfamily domain containing 2B; major facilitator superfamily domain-containing protein 2B; MFSD2B; RGD1561716 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388931, 500624 | |
| 4°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| A6NFX1 | |
| MFSD2B | |
| A 15 amino acid peptide near the center of human MFSD2B. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction