Learn More
DiscoverX MG132
Description
MG132 is a synthetic inhibitor that potently, selectively, and reversibly inhibits the proteasome, assisting in ROS formation and apoptosis. Also activates JNK1, and inhibits calpain. Provided as 5mg of dry powder.
Specifications
Specifications
| Quantity | 5 mg |