Learn More
DiscoverX MG132
Product Code. 17382429
Encompass
Change view
Click to view available options
:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17382429 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17382429 Supplier DiscoverX Supplier No. 921098
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MG132 is an inhibitor of the proteasome and calpain, and activates JNK1. Provided as 5mg of dry powder.
MG132 is a synthetic inhibitor that potently, selectively, and reversibly inhibits the proteasome, assisting in ROS formation and apoptosis. Also activates JNK1, and inhibits calpain. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg