missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGARP Monoclonal Antibody (OTI3D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811799S
This item is not returnable.
View return policy
Description
MGARP Monoclonal antibody specifically detects MGARP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MGARP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WYQ3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 400916 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D9 | |
| Unconjugated | |
| CHCHD10 | |
| C4orf49; CESP1; CESP-1; Corneal endothelium-specific protein 1; GS3582; HUMMR; hypoxia up-regulated mitochondrial movement regulator protein; MGARP; mitochondria localized glutamic acid rich protein; mitochondria-localized glutamic acid-rich protein; OSAP; ovary-specific acidic protein; Protein MGARP | |
| Human recombinant protein fragment corresponding to amino acids 17-142 of human CHCHD10 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction