Learn More
Invitrogen™ MGAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121001
Description
Positive test controls include: Mouse liver, Mouse kidney, Mouse heart, Rat liver. The target is usually found in the following locations: Golgi apparatus membrane, Single-pass type II membrane protein. Immunogen sequence: GSAVTHIRQP DLSSIAVPPD HRKFQGYYKI ARHYRWALGQ VFRQFRFPAA VVVEDDLEVA PDFFEYFRAT YPLLKADPSL WCVSAWNDNG KEQMVDASRP ELLYRTDFFP GLGWLLLAEL WAELEPKWPK AFWDDWMRRP EQRQGRACIR PEISRTMTFG RKGVSHGQFF DQHLKFIKLN QQFVHFTQLD LSYLQREAYD RDFLARVYGA PQLQVEKVRT NDRKELGEVR VQYTGRDSFK AFAKALGVMD DLKSGVPRAG YRGIVTFQFR GRRVHLAPPL TWEGYDPSWN.
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1, 2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene.
Specifications
| MGAT1 | |
| Polyclonal | |
| Unconjugated | |
| MGAT1 | |
| alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; GGNT1; glcNAc-T I; GLCNAC-TI; GLCT1; GLYT1; Gnt1; GNT-1; GnTI; GNT-I; mannoside acetylglucosaminyltransferase 1; mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; MGAT; Mgat1; Mgat-1; MGAT1 gene for N-acetylglucosaminyltransferase; N-acetylglucosaminyltransferase I; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I; OTTHUMP00000161546 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17308, 4245, 81519 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26572, P27808, Q09325 | |
| MGAT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 156-445 of human MGAT1 (NP_001108092.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.