missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MGAT2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA597186

Product Code. 16520022

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PFSIQLYPNE FPGSDPRDCP RDLPKNAALK LGCINAEYPD SFGHYREAKF SQTKHHWWWK LHFVWERVKI LRDYAGLILF LEEDHYLAPD FYHVFKKMWK LKQQECPECD VLSLGTYSAS RSFYGMADKV DVKTWKSTEH NMGLALTRNA YQKLIECTDT FCTYDDYNWD WTLQYLTVSC LPKFWKVLVP QIPRIFHAGD CGMHHKKTCR PSTQSAQIES LLNNNKQYMF PETLTISEKF TVVAISPPRK NGGWGDIRDH ELCKSYRRLQ; Positive Samples: K-562, Mouse kidney, Rat kidney; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MGAT2
Polyclonal
Unconjugated
MGAT2
AA407964; alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase-like protein; beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; CDG2A; CDGS2; CH73-301C19.1; GlcNAc-T II; GLCNACTII; GNT2; GNT-II; Mannoside acetylglucosaminyltransferase 2; mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; mgat2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II; zgc:162268
Rabbit
Affinity chromatography
RUO
217664, 4247, 94273
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q09326, Q10469, Q921V5
MGAT2
Recombinant fusion protein containing a sequence corresponding to amino acids 178-447 of human MGAT2 (NP_0023991).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.