Learn More
Invitrogen™ MGAT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596944
Description
Immunogen sequence: GCTVDMLQAV YGLDGIRLRR RQYYTMPNFR QYENRTGHIL VQWSLGSPLH FAGWHCSWCF TPEGIYFKLV SAQNGDFPRW GDYEDKRDLN YIRGLIRTGG WFDGTQQEYP PADPSEHMYA PKYLLKNYDR FHYLLDNPYQ EPRSTAAGGW RHRGPEGRPP ARGKLDEAEV; Positive Samples: SW620, U-251MG, SKOV3; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. MGAT3 (N-acetylglucosaminyltransferase III) transfers a GlcNAc residue to the beta-linked mannose of the trimannosyl core of N-linked oligosaccharides and produces a bisecting GlcNAc. Expression of this gene may be controlled by a multiple-promoter system.
Specifications
| MGAT3 | |
| Polyclonal | |
| Unconjugated | |
| MGAT3 | |
| 1110038J12Rik; AI848272; Beta-1,4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase; GGNT3; GlcNAc-T III; GNT3; GNT-III; mannoside acetyl glucosaminyltransferase 3; mannoside acetylglucosaminyltransferase 3; mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; MGAT3; N-acetylglucosaminyltransferase III; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III; RP5-1104E15.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17309, 29582, 4248 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02527, Q09327, Q10470 | |
| MGAT3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 364-533 of human MGAT3 (NP_0024003). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.