missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MGAT4C Polyclonal Antibody
GREENER_CHOICE

Product Code. 16399504
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16399504 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16399504 Supplier Invitrogen™ Supplier No. PA582547

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YKGTENKLKD DDFEEESFDI PDNPPASLYT NMNVFENYEA SKAYSSVDEY FWGKPPSTGD VFVIVFENPI IIKKIKVNTG TEDRQNDILH HGALDVGENV MPSKQRRQCS TYLRLGEFKN GNFEMSGVNQ KIPFDIHCMR IYVTKTQ Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 93%.

Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage ('GnT-VI' activity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGAT4C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGAT4C
Gene Accession No. Q9UBM8
Gene Alias 9130411I17Rik; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; glcNAc-T IVc; GnT-IVc; GntIVh; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family member C; MGAT4 family, member C; MGAT4C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; RGD1307871; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog
Gene Symbols MGAT4C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MGAT4C. Recombinant protein control fragment (Product #RP-90519).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.