missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT4C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17841-1-AP-150UL
This item is not returnable.
View return policy
Description
Glycosyltransferase that participates in the transfer of N-acetylglucosamine (GlcNAc) to the core mannose residues of N- linked glycans. Catalyzes the formation of the GlcNAcbeta1-4 branch on the GlcNAcbeta1-2Manalpha1-3 arm of the core structure of N-linked glycans. Essential for the production of tri- and tetra-antennary N-linked sugar chains. Does not catalyze the transfer of GlcNAc to the Manalpha1-6 arm to form GlcNAcBeta1-4Manalpha1-6 linkage (′GnT-VI′ activity).Specifications
| MGAT4C | |
| Polyclonal | |
| Unconjugated | |
| MGAT4C | |
| GlcNAc T IVc, GnT IVc, GNTIVH, HGNT IV H, MGAT4C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25834, 299756, 67569 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBM8 | |
| Mgat4c | |
| MGAT4C Fusion Protein Ag12201 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction