Learn More
Gr-1/Ly-6G Antibody (1A8), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-81053AF350
Product Code. 18248188
  • € 583.00 / 0.10mL
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gr-1/Ly-6G
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
LY6G6D
LEW/N rats immunized with 1.5x10E8 Ly-6G-transfected EL4J cells
0.1 mL
Innate Immunity
58530
Mouse
Purified
Flow Cytometry
1A8
Flow Cytometry
C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Rabbit
Protein A purified
RUO
Primary
Recognises the murine GPI-anchored protein Ly6G, a marker of granulocyte differentiation, maturation and, together with Ly6C, a component of the Granulocyte Receptor-1 Antigen (Gr-1). Unlike another commonly used clone, RB6-8C5, 1A8 does not bind Ly6C.
Store at 4°C in the dark.
IgG κ