missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC12965 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808275
This item is not returnable.
View return policy
Description
MGC12965 functions in DNA binding.Specifications
| MGC12965 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| MGC12965 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100287896 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| LOC100287896 | |
| LOC100287896 | |
| Full length human recombit protein of human MGC12965 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction