missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC12965 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808275S
This item is not returnable.
View return policy
Description
MGC12965 Monoclonal antibody specifically detects MGC12965 in Human samples. It is validated for Western BlotSpecifications
| MGC12965 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| LOC100287896; MGC12965; uncharacterized LOC100287896; uncharacterized protein LOC100287896 | |
| Full length human recombinant protein of human MGC12965 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| LOC100287896 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100287896 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction