Learn More
MGC12965 Mouse anti-Human, Clone: OTI3A9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808344
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MGC12965 functions in DNA binding.Specifications
| MGC12965 | |
| Monoclonal | |
| Unconjugated | |
| LOC100287896 | |
| LOC100287896 | |
| Full length human recombit protein of human MGC12965 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| MGC12965 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100287896 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.