missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MGC16703 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | MGC16703 |
| Synonym | tubulin, alpha pseudogene |
| Gene Symbol | TUBA3FP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction