missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC20410 Antibody (PCRP-BATF2-2B9), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20913PCP
This item is not returnable.
View return policy
Description
MGC20410 Monoclonal antibody specifically detects MGC20410 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| MGC20410 | |
| Monoclonal | |
| PerCP | |
| basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
| Recombinant fragment (around aa5-140) of human MGC20410 (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PCRP-BATF2-2B9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 116071 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction