missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MGC87631 Polyclonal Antibody

Product Code. 15782731
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15782731 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782731 Supplier Invitrogen™ Supplier No. PA554806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PSQAEGGEGG VACGTVEQMT WLCSLPHAVG GGDGDHSSTG AVGGHPRGPG EYCHLHEQRV HHHIFARGKR KGKNHVSNVV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 29%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGC87631
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC144NL
Gene Accession No. Q6NUI1
Gene Alias CCDC144NL; coiled-coil domain containing 144 family, N-terminal like; putative coiled-coil domain-containing protein 144 N-terminal-like
Gene Symbols CCDC144NL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MGC87631. Recombinant protein control fragment (Product #RP-93564).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.