missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mgea5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121776
This item is not returnable.
View return policy
Description
Mgea5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
| Mgea5 | |
| Polyclonal | |
| Unconjugated | |
| Oga | |
| 2810009A20Rik; 4833427O07Rik; 5830447M11Rik; AA408215; beta-hexosaminidase; Beta-N-acetylglucosaminidase; beta-N-acetylhexosaminidase; Bifunctional protein NCOAT; Hexc; Hy5; hyaluronidase in meningioma; KIAA0679; MEA5; meningioma expressed antigen 5 (hyaluronidase); meningioma-expressed antigen 5; Mgea5; mKIAA0679; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; OGA; O-GLCNACASE; Protein O-GlcNAcase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10724, 154968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O60502, Q8VIJ5 | |
| MGEA5, Oga | |
| Recombinant protein Meningioma Expressed Antigen 5. The antigen corresponds to amino acid range 6-240 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction