missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGLL Monoclonal Antibody (OTI 2B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502973S
This item is not returnable.
View return policy
Description
MGLL Monoclonal antibody specifically detects MGLL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| MGLL | |
| Monoclonal | |
| 0.44 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P31321 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI 2B6 | |
| Unconjugated | |
| PRKAR1B | |
| AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase | |
| Full length human recombinant protein of human PRKAR1B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction