missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGLL Mouse anti-Human, Clone: OTI2G8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502890
This item is not returnable.
View return policy
Description
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.Specifications
| MGLL | |
| Monoclonal | |
| Unconjugated | |
| MGLL | |
| HU-K5, HUK5, MAGL, MGL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11343 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99685 | |
| MGLL | |
| Full length human recombit protein of human MGLL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction