missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-39033PCP
This item is not returnable.
View return policy
Description
mGluR1 Polyclonal antibody specifically detects mGluR1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| mGluR1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
| Synthetic peptide comprising residues 1159-1171 [SSVPSSPVSESVL] of the human mGluR1 protein. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 2911 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction