Learn More
Invitrogen™ mGluR1/mGluR5 Monoclonal Antibody (N75/3), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545582
Description
1 μg/mL of MA5-45582 was sufficient for detection of mGluR1/5 glutamate receptor in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 130kDa (both mGluR1 and mGluR5). This antibody was formerly sold as clone S75-3.
may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.
Specifications
| mGluR1/mGluR5 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P23385, P31424, P41594, P97772, Q13255, Q3UVX5 | |
| Grm1, Grm5 | |
| Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| N75/3 | |
| PerCP | |
| Grm5 | |
| 4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; Glu5R; glutamate metabotropic receptor 1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; Gm10828; GPRC1A; Gprc1e; Grm1; GRM5; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGLU1; mGlu5; mGluR1; mGluR5; mGluR5b; mGlurR1a; nmf373; PPP1R85; PPP1R86; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86; rcw; SCAR13; wobl | |
| Mouse | |
| Protein G | |
| RUO | |
| 108071, 14816, 24414, 24418, 2911, 2915 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.