Learn More
Invitrogen™ mGluR1/mGluR5 Monoclonal Antibody (N75/33), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545552
Product Code. 17866712
  • € 455.00 / 100µg
Description
1 μg/mL of MA5-45552 was sufficient for detection of mGluR1/5 glutamate receptor in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 130kDa (both mGluR1 and mGluR5). This antibody was formerly sold as clone S75-33.
may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mGluR1/mGluR5
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
P23385, P31424, P41594, P97772, Q13255, Q3UVX5
Grm1, Grm5
Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
N75/33
PerCP
Grm5
4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; Glu5R; glutamate metabotropic receptor 1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; Gm10828; GPRC1A; Gprc1e; Grm1; GRM5; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGLU1; mGlu5; mGluR1; mGluR5; mGluR5b; mGlurR1a; nmf373; PPP1R85; PPP1R86; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86; rcw; SCAR13; wobl
Mouse
Protein G
RUO
108071, 14816, 24414, 24418, 2911, 2915
4°C