missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR2 Antibody (6E4F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61834-0.025ml
This item is not returnable.
View return policy
Description
mGluR2 Monoclonal specifically detects mGluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| mGluR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog | |
| Mouse | |
| 95.6 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 6E4F6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRM2 | |
| Purified recombinant fragment of human GRM2 (AA: extra 414-558) expressed in E. Coli. | |
| Protein G purified | |
| Apoptosis, Cancer, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
| 2912 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction