missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mGluR2/mGluR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146113
This item is not returnable.
View return policy
Description
This antibody is predicted react to human and mouse based on sequence homology.
The mGluR proteins (metabotropic Glutamate Receptors) are members of the G-protein-coupled receptor family and are functionally and pharmacologically distinct from the GluR proteins (ionotropic Glutamate Receptors). The eight currently known mGluR proteins are mediated by two G-proteins with opposing regulation of adenylate cyclase pathways. The activities of mGluR1 and mGluR5 are mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current. The remainder of the eight sub-types of mGluR have an activity mediated by a G-protein that inhibits adenylate cyclase activity. mGluR-3, which may interact with GRASP, acts as a receptor for glutamate.
Specifications
| mGluR2/mGluR3 | |
| Polyclonal | |
| Unconjugated | |
| Grm2 | |
| 0710001G23Rik; 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member C; GLUR2; GLUR3; glutamate metabotropic receptor 2; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; glutamate receptor, metabotropic 3; Gprc1b; GPRC1C; GRM2; Grm3; M mGlu3; Metabotropic glutamate receptor 2; metabotropic glutamate receptor 3; mGlu2; mGlu3; mGluR2; MGLUR3; mGluR7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24415, 24416 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P31421, P31422 | |
| Grm2, Grm3 | |
| Synthetic peptide corresponding to residues 860-875 of the rat mGlurR2/3 protein. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction