missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS899
This item is not returnable.
View return policy
Description
mGluR3 Polyclonal antibody specifically detects mGluR3 in Human, Mouse, Rat, Porcine, Bovine, Equine, Hamster, Primate, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| mGluR3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 16 μg/mL, Immunocytochemistry 1:10 - 1:500 | |
| GLUR3, glutamate receptor, metabotropic 3, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human mGluR3. | |
| 0.05 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 2913 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction