missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5 Antibody (464818), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4514P
This item is not returnable.
View return policy
Description
mGluR5 Monoclonal antibody specifically detects mGluR5 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| mGluR5 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 464818 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
| Mouse myeloma cell line NS0-derived recombinant human mGluR5, Gln21-Ser509, Accession # P41594 | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2915 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction