missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR6 Antibody (4C11E12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61851
This item is not returnable.
View return policy
Description
mGluR6 Monoclonal specifically detects mGluR6 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| mGluR6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| CSNB1B, glutamate receptor, metabotropic 6, GPRC1Fmetabotropic glutamate receptor 6, mGlu6, mGluR6, MGLUR6DKFZp686H1993 | |
| Mouse | |
| 95.5 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4C11E12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRM6 | |
| Purified recombinant fragment of human GRM6 (AA: extra 480-585) expressed in E. Coli. | |
| Protein G purified | |
| Amino Acids Drugs and other small molecules, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
| 2916 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction