missing translation for 'onlineSavingsMsg'
Learn More

mGluR6 Antibody (7D9C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 203.00 - € 568.00

Specifications

Antigen mGluR6
Clone 7D9C9
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18035586
View Documents
Novus Biologicals
NBP2-61850
0.1 mL
€ 568.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18691508
View Documents
Novus Biologicals
NBP2-61850-0.025ml
0.025 mL
€ 203.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

mGluR6 Monoclonal specifically detects mGluR6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

mGluR6
1 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
CSNB1B, glutamate receptor, metabotropic 6, GPRC1Fmetabotropic glutamate receptor 6, mGlu6, mGluR6, MGLUR6DKFZp686H1993
GRM6
IgG1
Protein G purified
7D9C9
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Monoclonal
Purified
Amino Acids Drugs and other small molecules, GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
2916
Purified recombinant fragment of human GRM6 (AA: extra 480-585) expressed in E. Coli.
Primary
95.5 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.