missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT Antibody (MGMT/8364R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21104F
This item is not returnable.
View return policy
Description
MGMT Monoclonal antibody specifically detects MGMT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MGMT | |
| Monoclonal | |
| FITC | |
| 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
| Recombinant fragment (around aa1-100) of human MGMT (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
| 4255 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MGMT/8364R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction