Learn More
MGMT Monoclonal Antibody (OTI17H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809255S
Encompass_Preferred
 View more versions of this product
Product Code. 17501304
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
MGMT Monoclonal antibody specifically detects MGMT in Human samples. It is validated for Western Blot
Specifications
Specifications
MGMT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P40199
Mouse
Affinity chromatography
RUO
4680
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI17H4
Unconjugated
CEACAM6
0-6-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase; Agat; AGT; AI267024; methylated DNA protein cysteine methyltransferase; methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O(6)-alkylguanine-DNA alkyltransferase; O-6-alkylguanine-DNA alkyltransferase; O6-alkylguanine-DNA alkyltransferase; O-6-methylguan; O6-methylguanine-DNA methyltranferase; O-6-methylguanine-DNA methyltransferase; O6-methylguanine-DNA methyltransferase; O-6-methylguanine-DNA-alkyltransferase
Human recombinant protein fragment corresponding to amino acids 35-257 of human CEACAM6 produced in E.coli.
30 μL