Learn More
MGMT Mouse anti-Human, Clone: OTI17H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809255
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
O6-Methylguanine-DNA Methyltransferase (MGMT) is an important DNA repair protein involved in tumor cell resistance to the cytostatic activity of chemotherapeutic alkylating agents. This protein is also effective in protecting normal cells against the genotoxic and carcinogenic effects of DNA alkylation. The alkylating drug resistance is caused by MGMT′s ability to remove DNA alkyl groups introduced in the O6 position of guanine. MGMT is expressed in highly variable amounts, depending upon the cell and tissue type, species, and cellular growth characteristics. In addition, MGMT activity varies among groups of tumors and within a particular type of tumor.Specifications
| MGMT | |
| Monoclonal | |
| Unconjugated | |
| MGMT | |
| O-6-methylguanine-DNA methyltransferase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4255 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI17H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P16455 | |
| MGMT | |
| Full length human recombit protein of human MGMT produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.