missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGP Antibody (OTI11G6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72660AF647
This item is not returnable.
View return policy
Description
MGP Monoclonal specifically detects MGP in Human samples. It is validated for Western Blot.
Specifications
| MGP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MGP | |
| Full length recombinant protein of human MGP (NP_000891) produced in E.coli. | |
| 0.1 mL | |
| 4256.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI11G6 | |
| Western Blot | |
| Cell growth-inhibiting gene 36 protein, matrix Gla protein, MGLAP, NTI | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction