missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGP Monoclonal Antibody (OTI9C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806404S
This item is not returnable.
View return policy
Description
MGP Monoclonal antibody specifically detects MGP in Human samples. It is validated for Western BlotSpecifications
| MGP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08493 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4256 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI9C9 | |
| Unconjugated | |
| MGP | |
| cell growth-inhibiting gene 36 protein; GIG36; MATRIX; matrix gamma-carboxyglutamate (gla) protein; matrix Gla protein; Matrix Gla protein long form; Matrix Gla protein short form; Mglap; Mgp; MGP precursor; NTI; similar to matrix Gla protein | |
| Full length human recombinant protein of human MGP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction