Learn More
Goat anti-Mouse IgG1 Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-68502V
Product Code. 30230650
  • ONLINE EXCLUSIVE PRICE
    € 508.00
    € 479.85 / 0.50mL
    Save € 28.15
    5% Off
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1
Polyclonal
50mM Sodium Borate
Goat
Affinity purified
RUO
Mouse
Purified
ELISA
DyLight 405
IgG1, Igh-4, RGD1359539, VH7183
Mouse IgG1 paraproteins
0.5 mL
Secondary
Store at 4°C in the dark.