missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGRN1 Antibody (OTI3C3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72665FR
This item is not returnable.
View return policy
Description
MGRN1 Monoclonal specifically detects MGRN1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| MGRN1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| MGRN1 | |
| Full length recombinant protein of human MGRN1 (NP_056061) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Primate | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3C3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.- | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 23295.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction