missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101664
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MGST1.
The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress.
Specifications
| MGST1 | |
| Polyclonal | |
| Unconjugated | |
| MGST1 | |
| 1500002K10Rik; glutathione S-transferase; glutathione S-transferase 12; Gst; GST12; MGC14525; MGST; mGST1; MGST-I; Microsomal glutathione S-transferase 1; microsomal glutathione S-transferase 1-like protein; microsomal glutathione-S-transferase; microsomal GST-1; Microsomal GST-I | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4257 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10620 | |
| MGST1 | |
| A synthesized peptide derived from human MGST1(Accession P10620), corresponding to amino acid residues R41-L91. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction