missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (ER-HR52), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64952H
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| MHC Class I | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| HLA-E | |
| Adherent FI (CBAXBL) bone marrow stromal cells | |
| 0.125 mL | |
| Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3133 | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| ER-HR52 | |
| Immunohistochemistry, Immunohistochemistry-Frozen | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2d,p,q. Mouse strains with the haplotypes H-2f,r,s,w17,w23,w27 show weak reactivity with this antibody. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction