missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (MEM-E/02), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-304AF700
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MHC Class I | |
| Monoclonal | |
| Alexa Fluor 700 | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Recombinant human HLA-E denatured heavy chain. | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology | |
| 3133 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MEM-E/02 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction