missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Antibody (OX18), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-6405PCP
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal specifically detects MHC Class I in Rat samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| MHC Class I | |
| Monoclonal | |
| PerCP | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes a monomorphic determinant of rat MHC Class I (RT1A), expressed by all rat strains. However, quantitative measurements suggest that not all of the class I molecules are recognised. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| OX18 | |
| Flow Cytometry, Flow (Intracellular) | |
| HLA-E | |
| Rat spleen glycoproteins | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Major Histocompatibility Complexes | |
| 3133 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction