missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I RT1Ac Antibody (OX-27), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64951C
This item is not returnable.
View return policy
Description
MHC Class I RT1Ac Monoclonal specifically detects MHC Class I RT1Ac in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| MHC Class I RT1Ac | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-E | |
| PHA activated rat lymphocytes | |
| 0.25 mL | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| OX-27 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| HLA-E | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB100-64951 recognises a polymorphic determinant (c haplotype) of rat Class I MHC Antigen (RT-1A). | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction