missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II Antibody (A6-61.3) [FITC], Novus Biologicals™

Product Code. 30532887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532887 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532887 Supplier Novus Biologicals Supplier No. NBP345120F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse samples. It is validated for Radioimmune Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II
Applications Radioimmune Assays (RIA)
Classification Monoclonal
Clone A6-61.3
Conjugate FITC
Formulation PBS
Gene Alias class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Host Species Mouse
Immunogen This antibody was obtained from a fusion of the non-producing plasmacytoma Ag8.653 with spleen cells from ATL mice that had been immunized with ATH spleen cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.